Imprimer
Rakoff staining solution
Fournisseur: PanReac AppliChem
A8112.0015EA 0 EUR
APLIA8112.0015
Rakoff staining solution
Rakoff supravital method is a fast-dyeing technique for the assessment of the hormonal situation. Eosinophilia and cyanophilia of the epthelium can be distinguished.
Used for smears of vaginal and cervical epithelium.