Imprimer

Catalase assay kit

Fournisseur: Abnova
ABNOKA0884EA 666 EUR
ABNOKA0884
Catalase assay kit
Tests Tests enzymatiques
Catalase Assay Kit is used for measuring catalase activity.

  • Detection method: Colorimetric, fluorometric
  • Reactivity: Mammals
  • Sensitivity: ≤1 µU or less of catalase activity

Catalase (EC 1.11.1.6) is a ubiquitous antioxidant enzyme that is present in nearly all living organisms. It functions to catalyse the decomposition of hydrogen peroxide (H₂O₂) to water and oxygen. The Catalase Assay Kit provides a highly sensitive, simple, direct and HTS-ready assay for measuring Catalase activity in biological samples. In the assay, catalase first reacts with H₂O₂ to produce water and oxygen, the unconverted H₂O₂ reacts with OxiRed probe to produce a product, which can be measured at 570 nm (colorimetric method) or at Ex/Em = 535/587 nm (fluorometric method). Catalase activity is reversely proportional to the signal.

Information de livraison: Kit contains Catalase Assay Buffer (25 ml); OxiRed probe in DMSO (200 µl), HRP-lyophilised (1 vial), H₂O₂ 0,88 M (25 µl), Stop Solution (1 ml) and Catalase Positive Control (2 µl).
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR