Belgium
Order Entry
Anti-PITPNB Rabbit Polyclonal Antibody
  BWRLBS5858
Anti-PITPNB Rabbit Polyclonal Antibody
  BWRLBS5858
 
262,00 €
   
  1 * 1 EA

 

  • Type d'anticorps:
    Primaire
  • Symbole de l'antigène:
    PITPNB
  • Clonalité:
    Polyclonal
  • Hôte:
    Rabbit
  • Isotype:
  • Réactivité:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Taille:
    100 µg
  • Cdt:
    100 µG

 

 

Synthetic peptide, corresponding to amino acids 226-271 of Human PITPNB.

PITPNB pAb detects endogenous levels of PITPNB protein.

Type: Primary
Antigen: PITPNB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat