Anti-ASM Rabbit Polyclonal Antibody
BWRLBS60373
- Type d'anticorps:Primaire
- Symbole de l'antigène:ASM
- Clonalité:Polyclonal
- Hôte:Rabbit
- Isotype:
- Réactivité:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Taille:100 µg
- Cdt:100 µG
A synthetic peptide corresponding to residues in Human ASM.
ASM pAb detects endogenous levels of Acid sphingomyelinase protein.
Type: Primary
Antigen: ASM
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat