Anti-NLK Rabbit Polyclonal Antibody
BWRLBS7516
- Type d'anticorps:Primaire
- Symbole de l'antigène:NLK
- Clonalité:Polyclonal
- Hôte:Rabbit
- ImmunoChimie:Yes
- Isotype:
- Réactivité:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Taille:100 µg
- Cdt:100 µG
Recombinant full length Human NLK.
NLK pAb detects endogenous levels of NLK protein.
Type: Primary
Antigen: NLK
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat