Anti-ASK1 Rabbit Polyclonal Antibody [clone: T77]
BWRLBS1019
- Type d'anticorps:Primaire
- Symbole de l'antigène:ASK1
- Clonalité:Polyclonal
- Clone:T77
- Hôte:Rabbit
- ImmunoChimie:Yes
- Isotype:
- Réactivité:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Taille:100 µg
- Cdt:100 µG
Synthetic peptide, corresponding to amino acids 43-90 of Human ASK1.
ASK1 (T77) pAb detects endogenous levels of ASK1 protein
Type: Primary
Antigen: ASK1
Clonality: Polyclonal
Clone: T77
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat