Belgium
Order Entry
Anti-p19ARF Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  BOSSBS-0534R-FITC
 :  Bioss
Anti-p19ARF Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
  BOSSBS-0534R-FITC
 :  Bioss
 
900,00 €
   
  1 * 100 µl

 

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    Cyclin-dependent kinase inhibitor 2A
  • Symbole de l'antigène:
    p19ARF
  • Clonalité:
    Polyclonal
  • Conjugaison:
    FITC (Fluorescein Isothiocyanate)
  • Hôte:
    Rabbit
  • ImmunoChimie:
    Yes
  • Isotype:
    IgG
  • Réactivité:
    Rat,
    Mouse
  • Environmentally Preferable:
  • Adsorption croisée:
    No
  • Formulaire:
    liquid
  • Synonymes antigène:
    CDKN2|p16-INK4|MLM|INK4|P16|p19ARF|p16-INK4a|p16INK4a|p16|CDKN2A|p16INK4|p19|p14|p16INK4A|CDK4I|INK4a|p14ARF|CMM2|ARF|MTS1|TP16
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Température de stockage:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Température de transport:
    4°C
  • Purification:
    Purified by Protein A
  • Cdt:
    100 µl

 

 

P19ARF Capable of inducing cell cycle arrest in G1 and G2 phases. Acts as a tumor suppressor. Binds to MDM2 and blocks its nucleocytoplasmic shuttling by sequestering it in the nucleolus. This inhibits the oncogenic action of MDM2 by blocking MDM2-induced degradation of p53 and enhancing p53-dependent transactivation and apoptosis. Also induces G2 arrest and apoptosis in a p53-independent manner by preventing the activation of cyclin B1/CDC2 complexes. Binds to BCL6 and down-regulates BCL6-induced transcriptional repression. Binds to E2F1 and MYC and blocks their transcriptional activator activity but has no effect on MYC transcriptional repression. Binds to TOP1/TOPOI and stimulates its activity. This complex binds to rRNA gene promoters and may play a role in rRNA transcription and/or maturation. Interacts with NPM1/B23 and promotes its polyubiquitination and degradation, thus inhibiting rRNA processing. Interacts with UBE2I/UBC9 and enhances sumoylation of a number of its binding partners including MDM2 and E2F1. Binds to HUWE1 and represses its ubiquitin ligase activity. May play a role in controlling cell proliferation and apoptosis during mammary gland development.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: p19ARF
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat