Anti-CKLF Rabbit Polyclonal Antibody (Alexa Fluor® 488)
BOSSBS-2394R-A488
: Bioss
- Type d'anticorps:Primaire
- Nom de l'antigène:chemokine-like factor
- Symbole de l'antigène:CKLF
- Clonalité:Polyclonal
- Conjugaison:Alexa Fluor® 488
- Hôte:Rabbit
- ImmunoChimie:Yes
- Isotype:IgG
- Réactivité:Human,Rat,Mouse
- Environmentally Preferable:
- Adsorption croisée:No
- Formulaire:liquid
- ID de gène:51192
- Synonymes antigène:CKLF4|C32|CKLF1|HSPC224|CKLF3|CKLF2|UCK-1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Température de stockage:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Température de transport:4°C
- Immunogène:30-65/152
- Purification:Purified by Protein A
- Cdt:100 µl
May play an important role in inflammation and regeneration of skeletal muscle. Partly inhibited by interleukin 10. Isoform 1 has chemotactic response in rat monocytes, neutrophils, and lymphocytes.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CKLF
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype:
Reactivity: