Anti-FBXO32 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
BOSSBS-2591R-FITC
: Bioss
- Type d'anticorps:Primaire
- Nom de l'antigène:F-Box Protein 32
- Symbole de l'antigène:FBXO32
- Clonalité:Polyclonal
- Conjugaison:FITC (Fluorescein Isothiocyanate)
- Hôte:Rabbit
- ImmunoChimie:Yes
- Isotype:IgG
- Réactivité:Human,Rat,Mouse
- Environmentally Preferable:
- Adsorption croisée:No
- Formulaire:liquid
- ID de gène:114907
- Synonymes antigène:MAFbx|Fbx32
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Température de stockage:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Température de transport:4°C
- Purification:Purified by Protein A
- Cdt:100 µl
Substrate recognition component of a SCF (SKP1-CUL1-F-box protein) E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins. Probably recognizes and binds to phosphorylated target proteins during skeletal muscle atrophy. Recognizes TERF1.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: Fbx32
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat