- Type d'anticorps:Primaire
- Nom de l'antigène:Histone cluster 4 H4
- Symbole de l'antigène:HIST4H4
- Clonalité:Polyclonal
- Conjugaison:Alexa Fluor® 647
- Hôte:Rabbit
- ImmunoChimie:Yes
- Isotype:IgG
- Réactivité:Human,Rat,Mouse
- Environmentally Preferable:
- Adsorption croisée:No
- Formulaire:liquid
- ID de gène:121504
- Synonymes antigène:H4FJ|HIST1H4H|H4/I|H4/M|H4FI|H4/K|H4FA|H4/G|HIST2H4B|H4F2|H4/O|HIST1H4L|H4/D|H4FC|H4/E|H4/B|H4FE|HIST1H4J|H4FH|H4FM|H4FO|H4FB|HIST1H4K|H4/N|H4/p|H4/J|HIST2H4|H4/C|HIST1H4F|H4FG|HIST1H4I|H4/A|H4FK|H4/H|H4FN|H4FD|HIST2H4A
- Modification:acetyl-Lys5
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Température de stockage:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Température de transport:4°C
- Purification:Purified by Protein A
- Cdt:100 µl
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. This gene is intronless and encodes a member of the histone H4 family. Transcripts from this gene lack polyA tails; instead, they contain a palindromic termination element. [provided by RefSeq, Jul 2008].
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: HIST4H4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat