Order Entry
Couch covers, SCOTT® EXTRA
  KIMB7398
Couch covers, SCOTT® EXTRA
  KIMB7398
559,00 €
  1 * 12 ROU
  • Emballage:
    12 rolls of 140 sheets
  • Couleur:
    bleu
  • Désignation:
    SCOTT® EXTRA couch cover roll, 2 ply
  • L×l:
    380×510 mm
  • Seq:
    02
  • Environmentally Preferable:
  • Cdt:
    12 Roll
This two ply couch cover is strong, tear resistant and durable.
  • For use in busy areas
  • Can be used with a dispensing system to help control use and reduce wastage
Ideal for: protecting couches and maintaining high levels of hygiene in the healthcare environment.
Dispenser
  •  
    Seq
    Désignation
    Cdt
    Prix
  • 07
    Distributeur mural, 7056, blanc, pour rouleaux d'une largeur max. de 500 mm
    1
    433,89
    1 * 1 ST
    1 * 1 ST - 104,00 €
    104,00 €