Imprimer
Lancing device, Compact Eject
Fournisseur: Vitrex Medical
VITREX®
MODU369400EA 4.8 EUR
MODU369400
Lancing device, Compact Eject
Lancettes
Lancing device with push button ejection designed for sampling in blood glucose monitoring.
  • Variable penetration, simple and quick activation and reactivation
  • Simple non-touch lancet removal
  • Clear adjustable and clear fixed end caps designed for Alternate Site Testing (AST)
Suitable for use with VITREX SOFT and most other popular lancet brands
Conditionnement: Clear plastic sleeves.