Imprimer
Anemometer, telescopic rotating probe
Fournisseur: Testo
TEST0635.9535EA 547 EUR
TEST0635.9535
Anemometer, telescopic rotating probe
This IAQ (Indoor Air Quality) probe is mainly involves in measuring inside of ducts.
  • Includes with 890 mm long telescopic handle
It features thermal probes, vane measurement and built-in differential pressure probe for pitot tube measurement.
Certifications: Conforms to protection class IP54