Imprimer
Film sealing device, Fermant® 40 N
KWPA2015465EA 0 EUR
KWPA2015465
Film sealing device, Fermant® 40 N
Fermant® 40 N sealing device is characterised by being user friendly. The use of a digital time control allows continuous control of the impulse and contact time. This allows optimum adaption to the respective type of film used.
  • Constantly even contact pressure
  • Continuous adjustability of the sealing process
  • Integrated item counter
  • Control by electric foot switch
Sealing l×w: 400×4 mm