Imprimer
Bead dispenser for 96 wells microtiter or deep well plates
IBSA96-2-4-32 IBSA96-2-999 IBSA96-2-4-20 IBSA96-2-1-28 IBSA96-2-4-25 IBSA96-2-1-25 IBSA96-2-1-47
Bead dispenser for 96 wells microtiter or deep well plates
The 96-well bead dispenser is used to dispense bead in 96-well microtiter and deep well plates. Can dispense a wide range of solid quantities in a highly precise and repeatable fashion.
  • Simple, fast and reliable
  • High productivity improvement 60-150 x
  • Easy cleaning with 70% ethanol or soap
  • Sterilisable and not autoclavable
  • Ergonomic use as it removes tedious repetitive tasks
  • Custom made volume plates are available
Material: ABS+PC, nylon, RVS304, anodised aluminum
* Customised size
Information de livraison: Supplied with converter and flight case.