The new Corning Dissolvable Microcarriers are designed to simplify the cell harvest, separation, and concentration processes. All microcarriers arrive pre-sterilized and pre-coated with Corning Synthemax™ II to enhance cell attachment for optimal yield and viability.
- Dissolvable for easy separation of cells from microcarriers
- Optical clarity for visualizing cell morphology
- Sterility Assurance Level (SAL) of 10⁻²
- Pre-coated for enhanced cell attachment
- Nonpyrogenic
They are dissolved within 10 to 20 minutes, allowing for the easy collection of cells without the need for microcarrier separation. Corning Dissolvable Microcarriers offer a cell separation that is faster, gentler, and more convenient than that of standard commercially available microcarriers. Ideal for cell therapy applications, vaccine production and large-volume adherent cell culture scale-up and cell banking.
Bead size: 200 - 300 µm
Surface area: 5000 cm² /g
Information de livraison: Supplied as dry, sterile powder.