Imprimer
Tissue embedder, EC 350
Fournisseur: Klinipath
KPXXEC-350EA 11420 EUR
KPXXEC-350
Tissue embedder, EC 350
The EC 350 is a modular paraffin embedding center featuring innovative design as well as easy operation. Different possibilities of setting up this instrument offer a simple and flexible adaptation to the working methods in the respective lab.
  • Dispensing console for easy operation by perfect ergonomy and fast specimen processing
  • Energy-saving operation by automatic turning on and off via automatic calculation
  • Non-tiring working by improved operating comfort, heated working surfaces
The cryo console fits harmonically into the overall system, can be placed either on the right or left side of the dispensing console. It has a large cooling surface with temperatures down to -12 °C and a steady temperature distribution over the entire surface.
Cold plate can operate connected to the dispensing unit or as stand-alone instrument.