Imprimer
Tampon TAE 50x (Tris-acetate-EDTA), solution aqueuse 50x concentrée Electran® pour l'électrophorèse
444125DEA 234 EUR
VWRC444125D
Tampon TAE 50x (Tris-acetate-EDTA), solution aqueuse 50x concentrée Electran® pour l'électrophorèse
DNases (exo and endonucleases), RNases and proteases are not detected.
50x concentrated solution of 2 M Tris 50 mM EDTA and 1 M acetic acid pH 8,0 in distilled, deionised water.