Imprimer
Film applicators, automatic, compact
Fournisseur: Industrial Physics
SIMEAB3655EA 2790 EUR
SIMEAB3655 SIMEAB3650
Film applicators, automatic, compact
Entry level film applicator that accepts charts up to A4 size. Application speed can be set from 1 to 150 mm/s.
  • Graphical LCD display
  • Ideal for application of film on paints, battery, cosmetics, adhesives
Suitable for both wires wound rod applicators and block applicators. Operating is easy through the standard operating interface.
Dimension: 335×400×190 mm.