Imprimer
Film flow meters
Fournisseur: Horiba
HORI2024080289EA 0 EUR
HORI2024080289 HORI2024080288 HORI2024080287 HORI2024080286
Film flow meters
These precision volumetric tubes with a soap film detector allow to measure the flow of a high number of gases accurately.
  • Flow rate measurement with a soap film detection
  • LCD digital display with remote control by PC
  • SF-2U features an internal atmospheric pressure sensor
Suitable for gas flow rates using one of the four volumetric units (VP series).