Belgium
Order Entry
Supelco® Organic Reference Standard, Perfluoroundecanoic acid
Supelco® Organic Reference Standard, Perfluoroundecanoic acid
  80444-50MG
 :  Merck
 :  
undefined
Supelco® Organic Reference Standard, Perfluoroundecanoic acid
  80444-50MG
 :  Merck
 :  
 
59,80 €
   
  1 * 50 mg

 

  • Cdt:
    50 mg
  • Normes:
    Analytical standard
  • N ° CAS.:
    2058-94-8
  • Désignation:
    Perfluoroundecanoic acid
  • Environmentally Preferable:
  • Conditions de stockage:
    RT
  • Description de la classe de danger:
    H302 + H332 - H351 - H360D - H362 - H372
  • Conditions de transport:
    STD
  • MDL No.:
    MFCD00153268
  • M-Clarity quality level:
    MQ100
  • Synonyme(s):
    PFAS; Heneicosafluoroundecanoic acid; PFUnDA
  • Point de fusion:
    96 - 101 °C (lit.)
  • Formula:
    CF₃(CF₂)₉CO₂H
  • Molecular weight:
    564,09
  • Point d'éclair:
    113 °C - Closed cup
  • Point d'ébullition:
    160 °C/60 mmHg (lit.)