Supelco® PESTANAL® Organic Reference Standard, τ-Fluvalinate
46294-100MG
: Merck
:
- Cdt:100 mg
- Normes:PESTANAL®, analytical standard
- N ° CAS.:102851-06-9
- Désignation:τ-Fluvalinate
- Environmentally Preferable:
- Conditions de stockage:2…8 °C
- Description de la classe de danger:H301 - H315 - H410
- Conditions de transport:STD
- MDL No.:MFCD00871307
- M-Clarity quality level:MQ100
- Synonyme(s):N-[2-Chloro-4-(trifluoromethyl)phenyl]-ᴅ-valine (RS)-cyano(3-phenoxyphenyl)methyl ester
- Molecular weight:502,91
- Point d'éclair:90 °C