RABS/isolator gloves, BioClean™ Clean Nitrile CGL
: Nitritex
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Catalog No:
- NITRCGL20NIT59
- NITRCGL33NIT59
- NITRCGL10NIT59
- NITRCGL36NIT59
- Modèle:
- CGL
- CGL
- CGL
- CGL
- Ambidextre/anatomique:
- Ambidextres
- Ambidextres
- Ambidextres
- Ambidextres
- Latex/Sans latex:
- Sans latex
- Sans latex
- Sans latex
- Sans latex
- Classe de la salle propre:
- 10/ISO 4; EU GMP (A)
- 10/ISO 4; EU GMP (A)
- 10/ISO 4; EU GMP (A)
- 10/ISO 4; EU GMP (A)
- Matière:
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Finition:
- Smooth
- Smooth
- Smooth
- Smooth
- Style de manchette:
- Roulé
- Roulé
- Roulé
- Roulé
- Norme EN:
- EN 388:2016 (4101X), EN ISO 374-1 Type A (AJKNOT), EN ISO 374-5 VIRUS, EN 421
- EN 388:2016 (4101X), EN ISO 374-1 Type A (AJKNOT), EN ISO 374-5 VIRUS, EN 421
- EN 388:2016 (4101X), EN ISO 374-1 Type A (AJKNOT), EN ISO 374-5 VIRUS, EN 421
- EN 388:2016 (4101X), EN ISO 374-1 Type A (AJKNOT), EN ISO 374-5 VIRUS, EN 421
- SeqDésignationTaille du gantCouleurLongueurCdt
- NITRCGL20NIT59CGL20NIT5901Taille du port 8 - 10" / 203 - 254 mm, manchette standard9,75blanc840 mm201 * 20 ST6 260,00 €
Specifications
ModèleCGLAmbidextre/anatomiqueAmbidextresEnvironmentally PreferableLatex/Sans latexSans latexClasse de la salle propre10/ISO 4; EU GMP (A)MatièreNitrileFinitionSmoothStyle de manchetteRouléNorme ENEN 388:2016 (4101X), EN ISO 374-1 Type A (AJKNOT), EN ISO 374-5 VIRUS, EN 421 - CGL10NIT5902Taille du port 10 - 12" / 254 - 305 mm, manchette épaisse9,75blanc840 mm201 * 20 ST5 730,00 €
Specifications
ModèleCGLAmbidextre/anatomiqueAmbidextresEnvironmentally PreferableLatex/Sans latexSans latexClasse de la salle propre10/ISO 4; EU GMP (A)MatièreNitrileFinitionSmoothStyle de manchetteRouléNorme ENEN 388:2016 (4101X), EN ISO 374-1 Type A (AJKNOT), EN ISO 374-5 VIRUS, EN 421 - NITRCGL33NIT59CGL33NIT5903Port size 10 - 12" / 254 - 305 mm, standard cuff9,75blanc840 mm11 * 1 ST
Specifications
ModèleCGLAmbidextre/anatomiqueAmbidextresEnvironmentally PreferableLatex/Sans latexSans latexClasse de la salle propre10/ISO 4; EU GMP (A)MatièreNitrileFinitionSmoothStyle de manchetteRouléNorme ENEN 388:2016 (4101X), EN ISO 374-1 Type A (AJKNOT), EN ISO 374-5 VIRUS, EN 421 - CGL36NIT5904Taille du port 12 - 14" / 305 - 356 mm, manchette standard9,75blanc840 mm201 * 20 ST6 990,00 €
Specifications
ModèleCGLAmbidextre/anatomiqueAmbidextresEnvironmentally PreferableLatex/Sans latexSans latexClasse de la salle propre10/ISO 4; EU GMP (A)MatièreNitrileFinitionSmoothStyle de manchetteRouléNorme ENEN 388:2016 (4101X), EN ISO 374-1 Type A (AJKNOT), EN ISO 374-5 VIRUS, EN 421