Order Entry
Iron ICP Standards, (CRMs), Fluka™
Iron ICP Standards, (CRMs), Fluka™
Iron ICP Standards, (CRMs), Fluka™
  • Environmentally Preferable:
  •  
    Élement
    Symbole
    Concentration
    Matrice
    Type de conditionnement
    Cdt
  • HONCE1106-100ML
    E1106-100ML
    Fer
    Fe
    1000 mg/L
    2% HNO₃
    Plastic bottle
    100 mL
    1 * 100 mL
    1 * 100 mL - 79,30 €
    79,30 €
    1 * 100 mL
       79,30 €
  • HONCE1021-100ML
    E1021-100ML
    Fer
    Fe
    1000 mg/L
    2% HCl
    Plastic bottle
    100 mL
    1 * 100 mL
    1 * 100 mL - 79,30 €
    79,30 €
    1 * 100 mL
       79,30 €
  • HONCE1148-100ML
    E1148-100ML
    Fer
    Fe
    10000 mg/L
    5% HCl
    Plastic bottle
    100 mL
    1 * 100 mL
    1 * 100 mL - 138,00 €
    138,00 €
    1 * 100 mL
       138,00 €
  • HONCE1218-100ML
    E1218-100ML
    Fer
    Fe
    10000 mg/L
    5% HNO₃
    Plastic bottle
    100 mL
    1 * 100 mL
    1 * 100 mL - 138,00 €
    138,00 €
    1 * 100 mL
       138,00 €