/
Belgium
Products
Services
Digital Solutions
Applications
Support
About
0,00 €
Order Entry
4-(Cyclopentylsulphanyl)phenol 95%
APOSOR957592-1G
:
Apollo Scientific
:
4-(Cyclopentylsulphanyl)phenol 95%
APOSOR957592-1G
:
Apollo Scientific
:
632,00 €
1 * 1 g
-
+
Type de conditionnement:
Bottle
Cdt:
1 g
Seq:
3047
Environmentally Preferable:
:
:
194.293 g/mol
:
Température ambiante
:
C₁₁H₁₄OS
:
MFCD12780261