Order Entry
VWR®, Septa 16 mm
undefined
VWR®, Septa 16 mm
  •  
    Seq
    Ø
    Septum
    Dureté
    Épaisseur
    Cdt
  • 548-0537A
    01
    16 mm
    Red PTFE/white silicone/red PTFE
    45° shore A
    1,0 mm
    1.000
    1 * 1.000 ST
    1 * 1.000 ST - 703,00 €
    703,00 €
    1 * 1.000 ST
       703,00 €
    Specifications
    Environmentally Preferable
  • 548-0533A
    02
    16 mm
    Red-orange natural rubber/transparent TEF
    60° shore A
    1,3 mm
    1.000
    1 * 1.000 ST
    1 * 1.000 ST - 204,00 €
    204,00 €
    1 * 1.000 ST
       204,00 €
    Specifications
    Environmentally Preferable
  • 548-0534A
    03
    16 mm
    White silicone/red PTFE
    45° shore A
    1,3 mm
    1.000
    1 * 1.000 ST
    1 * 1.000 ST - 521,00 €
    521,00 €
    1 * 1.000 ST
       521,00 €
    Specifications
    Environmentally Preferable
  • 548-0795A
    04
    16 mm
    Red butyl/grey PTFE
    50° shore A
    1,3 mm
    1.000
    1 * 1.000 ST
    1 * 1.000 ST - 268,00 €
    268,00 €
    1 * 1.000 ST
       268,00 €
    Specifications
    Environmentally Preferable
  • 548-1063A
    05
    16 mm
    White silicone/red PTFE
    55° shore A
    1,5 mm
    1.000
    1 * 1.000 ST
    1 * 1.000 ST - 509,00 €
    509,00 €
    1 * 1.000 ST
       509,00 €
    Specifications
    Environmentally Preferable
  • 548-0539A
    06
    16 mm
    Red butyl/grey PTFE
    50° shore A
    1,6 mm
    1.000
    1 * 1.000 ST
    1 * 1.000 ST - 356,00 €
    356,00 €
    1 * 1.000 ST
       356,00 €
    Specifications
    Environmentally Preferable
  • 548-0538A
    07
    16 mm
    Transparent blue silicone/white PTFE
    45° shore A
    1,7 mm
    1.000
    1 * 1.000 ST
    1 * 1.000 ST - 598,00 €
    598,00 €
    1 * 1.000 ST
       598,00 €
    Specifications
    Environmentally Preferable
  • 548-0531A
    08
    16 mm
    Natural silicone/beige PTFE
    50° shore A
    1,9 mm
    1.000
    1 * 1.000 ST
    1 * 1.000 ST - 505,00 €
    505,00 €
    1 * 1.000 ST
       505,00 €
    Specifications
    Environmentally Preferable
  • 548-0540A
    09
    16 mm
    Red butyl/grey PTFE
    50° shore A
    2,0 mm
    1.000
    1 * 1.000 ST
    1 * 1.000 ST - 360,00 €
    360,00 €
    1 * 1.000 ST
       360,00 €
    Specifications
    Environmentally Preferable
  • 548-0864A
    10
    16 mm
    Beige silicone/grey PTFE
    45° shore A
    2,5 mm
    1.000
    1 * 1.000 ST
    1 * 1.000 ST - 602,00 €
    602,00 €
    1 * 1.000 ST
       602,00 €
    Specifications
    Environmentally Preferable
  • 548-1832A
    11
    16 mm
    Transparent blue silicone/white PTFE
    45° shore A
    3,0 mm
    1.000
    1 * 1.000 ST
    1 * 1.000 ST - 856,00 €
    856,00 €
    1 * 1.000 ST
       856,00 €
    Specifications
    Environmentally Preferable
  • 548-2358A
    12
    16 mm
    Transparent blue silicone/white PTFE
    45° shore A
    3,0 mm
    1.000
    1 * 1.000 ST
    1 * 1.000 ST - 563,00 €
    563,00 €
    1 * 1.000 ST
       563,00 €
    Specifications
    Environmentally Preferable
  • 548-0532A
    13
    16 mm
    Cream silicone/beige PTFE
    50° shore A
    3,2 mm
    100
    1 * 100 ST
    1 * 100 ST - 59,00 €
    59,00 €
    1 * 100 ST
       59,00 €
    Specifications
    Environmentally Preferable