Order Entry
Media IDMEM W/GLN+HPS power, Corning®
  50-016-PB
 :  Corning
 :  
undefined
Media IDMEM W/GLN+HPS power, Corning®
  50-016-PB
 :  Corning
 :  
129,76 €
  1 * 1 ST
  • Format du milieu:
    Powder
  • Cdt:
    1
  • Désignation:
    Iscove’s Modified DMEM: with L-glutamine and 25 mM HEPES, without [beta]-thioglycerol, [beta]-mercaptoethanol, and sodium bicarbonate
  • Environmentally Preferable: