Order Entry
CPC® Sanitary Steam-in-Place Connectors, Two-Cycle
CPC® Sanitary Steam-in-Place Connectors, Two-Cycle
undefined
CPC® Sanitary Steam-in-Place Connectors, Two-Cycle
  • Max. operating pressure:
  • 20 psi
  • 20 psi
  • 20 psi
  • Max. temperature:
  • 40 °C (104 °F)
  • 40 °C (104 °F)
  • 40 °C (104 °F)
  • Température min.:
  • 4 °C (39 °F)
  • 4 °C (39 °F)
  • 4 °C (39 °F)
  • Seal material:
  • Platinum-cured Silicone
  • Platinum-cured Silicone
  • Platinum-cured Silicone
  • Sterilisation:
  • Gamma or steam-in-place (SIP)
  • Gamma or steam-in-place (SIP)
  • Gamma or steam-in-place (SIP)
  • Garantie:
  • 1 year
  • 1 year
  • 1 year
Bulk product
  •  
    Désignation
    Barb size/tubing int.Ø (port 3)
    Type de connection
    Fitting material
    Flow pattern
    Sanitary clamp size (Port 1)
    Sanitary clamp size (Port 2)
  • 31309-62BLK
    Sanitary Steam-in-place connector, two-cycle, ¹/₂ID׳/₄×1½" Sanitary, STC2020200; 5/PK
    ¹/₂"
    Sanitary to Barb
    Polysulfone
    Straight
    1-½"
    ¾"
    1 * 5 ST
    1 * 5 ST - 971,00 €
    971,00 €
    1 * 5 ST
       971,00 €
    Specifications
    Environmentally Preferable
    Max. operating pressure
    20 psi
    Max. temperature
    40 °C (104 °F)
    Température min.
    4 °C (39 °F)
    Seal material
    Platinum-cured Silicone
    Sterilisation
    Gamma or steam-in-place (SIP)
    Garantie
    1 year
  • 31604-36BLK
    Sanitary Steam-in-Place Connector, Two-Cycle, ¹/₂" ID׳/₄"׳/₄" Sanitary, STC2020000; 5/PK
    ½"
    Sanitary to Barb
    Polysulfone
    Straight
    ¾"
    ¾"
    1 * 5 ST
    1 * 5 ST - 1 040,00 €
    1 040,00 €
    1 * 5 ST
       1 040,00 €
    Specifications
    Environmentally Preferable
    Max. operating pressure
    20 psi
    Max. temperature
    40 °C (104 °F)
    Température min.
    4 °C (39 °F)
    Seal material
    Platinum-cured Silicone
    Sterilisation
    Gamma or steam-in-place (SIP)
    Garantie
    1 year
Down pack product
  •  
    Désignation
    Barb size/tubing int.Ø (port 3)
    Type de connection
    Fitting material
    Flow pattern
    Sanitary clamp size (Port 1)
    Sanitary clamp size (Port 2)
  • 31604-36
    Sanitary Steam-in-Place Connector, Two-Cycle, ¹/₂" ID׳/₄"׳/₄" Sanitary, STC2020000; 1/PK
    ½"
    Sanitary to Barb
    Polysulfone
    Straight
    ¾"
    ¾"
    1 * 1 ST
    1 * 1 ST - 376,00 €
    376,00 €
    1 * 1 ST
       376,00 €
    Specifications
    Environmentally Preferable
    Max. operating pressure
    20 psi
    Max. temperature
    40 °C (104 °F)
    Température min.
    4 °C (39 °F)
    Seal material
    Platinum-cured Silicone
    Sterilisation
    Gamma or steam-in-place (SIP)
    Garantie
    1 year