Order Entry
Parker Hannifin Domnick Hunter Air Purification Systems
undefined
Parker Hannifin Domnick Hunter Air Purification Systems
  • Pression max.:
  • 20 bar (290 psi)
  • 16 bar (232 psi)
  • 16 bar (232 psi)
  • 16 bar (232 psi)
  • 16 bar (232 psi)
  • 16 bar (232 psi)
  • Residual oil content:
  • 0,003 ppm at 70 °F (0,003 mg/m³ at 21 °C)
  • 0,5 ppm at 70 °F (0,6 mg/m³ at 21 °C)
  • 0,5 ppm at 70 °F (0,6 mg/m³ at 21 °C)
  • 0,1 ppm at 70 °F (0,01 mg/m³ at 21 °C)
  • 0,1 ppm at 70 °F (0,01 mg/m³ at 21 °C)
  • 0,1 ppm at 70 °F (0,01 mg/m³ at 21 °C)
  • Ø:
  • 9,65 cm (3,8")
  • 9,65 cm (3,8")
  • 12,95 cm (5,1")
  • 7,62 cm (3")
  • 9,65 cm (3,8")
  • 9,65 cm (3,8")
  • Garantie:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  •  
    Désignation
    Hauteur
    Matière
    Filter type
    Micron nominal
    Flow range
    Max. temp.
  • 29587-23
    Air Purification System Replacement Filter Element, Activated Carbon Filter, 42 ft.³/min
    236,2 mm
    Deep-bed activated carbon
    Activated carbon
    0,003 µm
    42 ft.³/min at 100 psig
    30 °C
    1 * 1 ST
    1 * 1 ST - 153,00 €
    153,00 €
    1 * 1 ST
       153,00 €
    Specifications
    Environmentally Preferable
    Pression max.
    20 bar (290 psi)
    Residual oil content
    0,003 ppm at 70 °F (0,003 mg/m³ at 21 °C)
    Ø
    9,65 cm (3,8")
    Garantie
    1 year
  • 29587-03
    Air Purification System Replacement Filter Element, Prefilter, 42 ft.³/min
    236,2 mm
    Deep-bed pleated oleophobic nanofiber
    Prefilter
    1 µm
    42 ft.³/min at 100 psig
    80 °C
    1 * 1 ST
    1 * 1 ST - 164,00 €
    164,00 €
    1 * 1 ST
       164,00 €
    Specifications
    Environmentally Preferable
    Pression max.
    16 bar (232 psi)
    Residual oil content
    0,5 ppm at 70 °F (0,6 mg/m³ at 21 °C)
    Ø
    9,65 cm (3,8")
    Garantie
    1 year
  • 29587-07
    Air Purification System Replacement Filter Element, Prefilter, 127 ft.³/min
    274,3 mm
    Deep-bed pleated oleophobic nanofiber
    Prefilter
    1 µm
    127 ft.³/min at 100 psig
    80 °C
    1 * 1 ST
    1 * 1 ST - 269,00 €
    269,00 €
    1 * 1 ST
       269,00 €
    Specifications
    Environmentally Preferable
    Pression max.
    16 bar (232 psi)
    Residual oil content
    0,5 ppm at 70 °F (0,6 mg/m³ at 21 °C)
    Ø
    12,95 cm (5,1")
    Garantie
    1 year
  • 29587-11
    Air Purification System Replacement Filter Element, Microfilter, 21 ft.³/min
    182,9 mm
    Deep-bed pleated oleophobic nanofiber
    Microfilter
    0,01 µm
    21 ft.³/min at 100 psig
    80 °C
    1 * 1 ST
    1 * 1 ST - 129,00 €
    129,00 €
    1 * 1 ST
       129,00 €
    Specifications
    Environmentally Preferable
    Pression max.
    16 bar (232 psi)
    Residual oil content
    0,1 ppm at 70 °F (0,01 mg/m³ at 21 °C)
    Ø
    7,62 cm (3")
    Garantie
    1 year
  • 29587-13
    Air Purification System Replacement Filter Element, Microfilter, 42 ft.³/min
    236,2 mm
    Deep-bed pleated oleophobic nanofiber
    Microfilter
    0,01 µm
    42 ft.³/min at 100 psig
    80 °C
    1 * 1 ST
    1 * 1 ST - 164,00 €
    164,00 €
    1 * 1 ST
       164,00 €
    Specifications
    Environmentally Preferable
    Pression max.
    16 bar (232 psi)
    Residual oil content
    0,1 ppm at 70 °F (0,01 mg/m³ at 21 °C)
    Ø
    9,65 cm (3,8")
    Garantie
    1 year
  • 29587-15
    Air Purification System Replacement Filter Element, Microfilter, 64 ft.³/min
    236,2 mm
    Deep-bed pleated oleophobic nanofiber
    Microfilter
    0,01 µm
    64 ft.³/min at 100 psig
    80 °C
    1 * 1 ST
    1 * 1 ST - 247,00 €
    247,00 €
    1 * 1 ST
       247,00 €
    Specifications
    Environmentally Preferable
    Pression max.
    16 bar (232 psi)
    Residual oil content
    0,1 ppm at 70 °F (0,01 mg/m³ at 21 °C)
    Ø
    9,65 cm (3,8")
    Garantie
    1 year