Masterflex® Transfer Tubing, Tygon® ND-100-80 Microbore, Avantor®
Fournisseur: Avantor Fluid Handling
Certificats
À propos de cet article
Clear formulation for easier sample viewing.
- Ideal for general-purpose, single-use, and extended fluid path applications
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimise downtime
- Regulatory approvals to match your unique application
Formulation: Tygon ND-100-80
Applications: General laboratory and pharmaceutical use, especially with precision injection or dispensing applications. Also excellent with low-flow peristaltic pump applications.
Characteristics: Extremely flexible. Non-toxic; nonpyrogenic; biocompatible. Crystal clear.
Certifications: Resin meets USP Class VI requirements.
Temperature range: -25 to 185 °F (-31 to 85 °C)
Sterilize: Autoclave, ethylene oxide, or gamma irradiation.
- Environmentally Preferable:
Spécifications
- Clarity:
- Translucent
- Translucent
- Translucent
- Translucent
- Translucent
- Length per pack:
- 30,5 m (100 ft.)
- 30,5 m (100 ft.)
- 30,5 m (100 ft.)
- 30,5 m (100 ft.)
- 30,5 m (100 ft.)
- Max. temperature:
- 85 °C (185 °F)
- 85 °C (185 °F)
- 85 °C (185 °F)
- 85 °C (185 °F)
- 85 °C (185 °F)
- Température min.:
- –31 °C (–25 °F)
- –31 °C (–25 °F)
- –31 °C (–25 °F)
- –31 °C (–25 °F)
- –31 °C (–25 °F)
- Regulatory approvals:
- Non-DEHP;USP
- Non-DEHP;USP
- Non-DEHP;USP
- Non-DEHP;USP
- Non-DEHP;USP
- Sterilisation method autoclave:
- Yes
- Yes
- Yes
- Yes
- Yes
- Sterilisation method EtO:
- Yes
- Yes
- Yes
- Yes
- Yes
- Sterilisation method gamma:
- Yes
- Yes
- Yes
- Yes
- Yes
Spécifications
Informations sur le produit
- RéférenceDésignationTube Ø int.Épaisseur de paroiTube Ø ext.Matériau de tuyauxLongueurCouleurCdtDisponibilitéPrix
- Microbore Transfer Tubing, Tygon® ND-100-80, 0,010" ID x 0,030" OD; 100 Ft0,25 mm (0,01")0,25 mm0,76 mm (0,03")Tygon® ND-100-8030,5 mtranslucide100 Feet1 * 100 Voet98,50 €
Specifications
Environmentally PreferableClarityTranslucentLength per pack30,5 m (100 ft.)Max. temperature85 °C (185 °F)Température min.–31 °C (–25 °F)Regulatory approvalsNon-DEHP;USPSterilisation method autoclaveYesSterilisation method EtOYesSterilisation method gammaYes - Microbore Transfer Tubing, Tygon® ND-100-80, 0,050" ID x 0,090" OD; 100 Ft1,27 mm (0,05")0,508 mm2,29 mm (0,09")Tygon® ND-100-8030,5 mtranslucide100 Feet1 * 100 Voet119,00 €
Specifications
Environmentally PreferableClarityTranslucentLength per pack30,5 m (100 ft.)Max. temperature85 °C (185 °F)Température min.–31 °C (–25 °F)Regulatory approvalsNon-DEHP;USPSterilisation method autoclaveYesSterilisation method EtOYesSterilisation method gammaYes - Microbore Transfer Tubing, Tygon® ND-100-80, 0,030" ID x 0,090" OD; 100 Ft0,76 mm (0,03")0,76 mm2,29 mm (0,09")Tygon® ND-100-8030,5 mtranslucide100 Feet1 * 100 Voet119,00 €
Specifications
Environmentally PreferableClarityTranslucentLength per pack30,5 m (100 ft.)Max. temperature85 °C (185 °F)Température min.–31 °C (–25 °F)Regulatory approvalsNon-DEHP;USPSterilisation method autoclaveYesSterilisation method EtOYesSterilisation method gammaYes - Microbore Transfer Tubing, Tygon® ND-100-80, 0,020" ID x 0,060" OD; 100 Ft0,51 mm (0,02")0,508 mm1,52 mm (0,06")Tygon® ND-100-8030,5 mtranslucide100 Feet1 * 100 Voet101,00 €
Specifications
Environmentally PreferableClarityTranslucentLength per pack30,5 m (100 ft.)Max. temperature85 °C (185 °F)Température min.–31 °C (–25 °F)Regulatory approvalsNon-DEHP;USPSterilisation method autoclaveYesSterilisation method EtOYesSterilisation method gammaYes - Microbore Transfer Tubing, Tygon® ND-100-80, 0,040" ID x 0,070" OD; 100 Ft1,02 mm (0,04")0,38 mm1,78 mm (0,07")Tygon® ND-100-8030,5 mtranslucide100 Feet1 * 100 Voet127,00 €
Specifications
Environmentally PreferableClarityTranslucentLength per pack30,5 m (100 ft.)Max. temperature85 °C (185 °F)Température min.–31 °C (–25 °F)Regulatory approvalsNon-DEHP;USPSterilisation method autoclaveYesSterilisation method EtOYesSterilisation method gammaYes