VWR®, KED Setup Solution, 2 components: Co 10 ug/l, Ce 1 ug/l in 1% HCl
Supplier: Avantor
VWR®
91173.230EA 161 EUR
91173.230 91173.180
VWR®, KED Setup Solution, 2 components: Co 10 ug/l, Ce 1 ug/l in 1% HCl
Standards Cationic Standards Multi-Element Standards
Certifications: ISO 17025/17034