VWR®, Quality Control Standard, Pure (Pure XVI), 21 Elements: 100 mg/l each of As, Be, Ca, Cd, Co, Cr, Cu, Fe, Li, Mg, Mn, Mo, Ni, Pb, Sb, Se, Sr, Ti, Tl, V, Zn in 5% HNO₃
Supplier: VWR Chemicals
91194.180EA 366 EUR
91194.180
VWR®, Quality Control Standard, Pure (Pure XVI), 21 Elements: 100 mg/l each of As, Be, Ca, Cd, Co, Cr, Cu, Fe, Li, Mg, Mn, Mo, Ni, Pb, Sb, Se, Sr, Ti, Tl, V, Zn in 5% HNO₃
Standards Cationic Standards Multi-Element Standards