VWR®, ICP-MS Stock Tuning Solution, 5 components: 10 mg/l each of Li, Y, Ce, Co, Tl in 2% HNO₃
Supplier: Avantor
VWR®
91204.180EA 203 EUR
91204.180
VWR®, ICP-MS Stock Tuning Solution, 5 components: 10 mg/l each of Li, Y, Ce, Co, Tl in 2% HNO₃
Standards Cationic Standards Multi-Element Standards
Certifications: ISO 17025/17034