VWR®, Standard for determination of 4 main elements: 5000 mg/l each of Ca, K, Mg, Na in 5% HNO₃
Supplier: VWR Chemicals
91284.260EA 376 EUR
91284.260 91284.180
VWR®, Standard for determination of 4 main elements: 5000 mg/l each of Ca, K, Mg, Na in 5% HNO₃
Standards Cationic Standards Multi-Element Standards
Certifications: ISO 17025/17034