Silicone keyboards for the medical sector, GCQ CleanType® Easy Protect Compact
Supplier: GETT GERÄTETECHNIK GMBH
KG29213 KG29215
GETTKG29213EA 95.5 EUR
GETTKG29213 GETTKG29215
Silicone keyboards for the medical sector, GCQ CleanType® Easy Protect Compact
Keyboards
The CleanType® Easy Protect Compact is a silicone keyboard that is easy to clean thanks to its water and dust resistance, making it ideal for use in hygiene sensitive areas such as hospitals, clinics, medical practices, laboratories, or pharmaceutical companies.
  • Medical-grade silicone keyboard with IP 68 protection rating; perfect for use in hygiene sensitive environments
  • 15 cm cable with waterproof USB silicone cap plus USB extension cable
  • Completely washable and disinfectable; can be cleaned with common cleaning agents
  • KeyLock function to switch off the keyboard during cleaning
  • 3 status LEDs (scroll, caps lock, KeyLock)
  • Two year warranty
  • Engineered in Germany
The CleanType® Easy Protect Compact silicone keyboard can be individualised by having a company logo printed on it, and therefore integrating it perfectly into a current corporate identity. In addition this keyboard can be equipped with any existing country layout.
Certifications: CE, FCC Part 15 Class B, REACH, IP 68.
Ordering information: Includes USB extension cable.
Packaging: Keyboards are safely and environmentally friendly packed in brown cardboard boxes.