Vertical electrophoresis system, MV10
Supplier: EDVOTEK EUROPE LTD
581
732-1427EA 379 EUR
732-1427
Vertical electrophoresis system, MV10
Electrophoresis Systems Vertical Gel Electrophoresis Systems
Designed for separation of proteins on polyacrylamide gels, the MV10 unit holds one 9×19 cm gel cassette and can accommodate most pre-cast or self-made gels.
  • Holds one gel cassette
  • All platinum electrodes
  • Safety interlock cover
  • Safety electrical leads