Industrial gloves, Polytril™ Skin
Supplier: HONEYWELL SAFETY
2232278/8
112-1637EA 39.3 EUR
112-1637
Industrial gloves, Polytril™ Skin
Gloves
The silicone-free polyamide knitted glove has an ultra thin foam nitrile coating on the palm and fingers, an open back and elasticated knitted wrists.
  • Ultra tight knitting gives perfect support, limits the penetration of dirt and offers good sensitivity
  • Thin nitrile coating provides oil and abrasion resistance and breathability
  • Non pilling, the polyamide is suitable for precision work
  • Black colour suitable for dirty applications and environments
Ideal for light handling and assembly in wet and greasy environments.
Gauge: 15
Length: 245 mm
Colour: Black
EN 388 (3131), Cat. III
Certifications: EN 388:2016 (3121X), Cat. III