2-(3-Fluorophenyl)ethanol 99%
Supplier: Thermo Scientific
Warning
304690250
ACRO304690250EA 589 EUR
ACRO304690250
2-(3-Fluorophenyl)ethanol 99%
2-(3-Fluorophenyl)ethanol
Formula: C₈H₉FO
MW: 140.16 g/mol
Boiling Pt: 90 °C (3 mmHg)
Density: 1.12 g/cm³
Flash Pt: 107 °C
Storage Temperature: Ambient
MDL Number: MFCD00045998
CAS Number: 52059-53-7
UN: 2810
ADR: 6.1,III
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5070 to 1.5100 (20°C, 589 nm)
GC ≥98.5 %