3-Iodobenzylamine 97%
Supplier: Thermo Scientific
Danger
Synonyms: (3-Iodophenyl)methanamine
367320010 367320050 367320200
ACRO367320010EA 117 EUR
ACRO367320010 ACRO367320050 ACRO367320200
3-Iodobenzylamine 97%
3-Iodobenzylamine
Formula: C₇H₈IN
MW: 233.05 g/mol
Boiling Pt: 132 °C (8 mmHg)
Density: 1.748 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00192227
CAS Number: 696-40-2
UN: 2735
ADR: 8,III
Order Now
Specification Test Results
Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6380 to 1.6400 (20°C, 589 nm)
GC ≥97.5 %