1-(2-(Trifluoromethyl)phenyl)imidazole ≥98% (by GC)

Synonyms: 1-(2-(Trifluoromethyl)phenyl)-1H-imidazole,TRIM

ALX270170M050
ALEXALX270170M050EA 43 EUR
ALEXALX270170M050
1-(2-(Trifluoromethyl)phenyl)imidazole ≥98% (by GC)
1-(2-(Trifluoromethyl)phenyl)imidazole

Potent neuronal nitric oxide synthase (nNOS; NOS I) inhibitor.


  • nNOS inhibitor


White solid.


Soluble in DMSO (30mg/ml) or dichloromethane (25%); slightly soluble in water (5mg/ml), 0.1 N HCl (6mg/ml) or 0.1 N NaOH (2mg/ml).

Formula: C₁₀H₇F₃N₂
MW: 212.17 g/mol
Boiling Pt: 75 °C (0.025 mmHg)
Melting Pt: 50…53 °C
MDL Number: MFCD00041206
CAS Number: 25371-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR