Tribenzylphosphine

Supplier: Thermo Scientific
Warning

10173.03 10173.06 10173.14
10173.03EA 63.3 EUR
10173.03 10173.06 10173.14
Tribenzylphosphine
Tribenzylphosphine
Formula: C₂₁H₂₁P
MW: 304.37 g/mol
Boiling Pt: 183…185 °C (0.2 mmHg)
Melting Pt: 93…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00014439
CAS Number: 7650-89-7
EINECS: 231-608-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR