3,3'-Dinitrobenzophenone 96%

Supplier: Thermo Scientific
Warning

Synonyms: Bis(3-nitrophenyl) ketone,Bis(3-nitrophenyl)methanone

B21153.06
B21153.06EA 53.2 EUR
B21153.06
3,3'-Dinitrobenzophenone 96%
3,3'-Dinitrobenzophenone
Formula: C₁₃H₈N₂O₅
MW: 272.22 g/mol
Melting Pt: 153…155 °C
Storage Temperature: Ambient
MDL Number: MFCD00039740
CAS Number: 21222-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR