3-(Aminomethyl)pyridine ≥99%

Supplier: Thermo Scientific
Danger

Synonyms: 3-PicolylaminePyridin-3-ylmethanamine(3-Pyridyl)methylamine3-Pyridylmethylamine3-Picolamine

B23634.14EA 38.6 EUR
B23634.14
3-(Aminomethyl)pyridine ≥99%
3-(Aminomethyl)pyridine
Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 226 °C
Melting Pt: –21 °C
Density: 1.062
Flash Pt: 100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006412
CAS Number: 3731-52-0
EINECS: 223-091-0
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR