4-Methyl-2-nitrophenyl isothiocyanate 95%

Supplier: Thermo Scientific
Danger

L10438.03EA 36.8 EUR
L10438.03
4-Methyl-2-nitrophenyl isothiocyanate 95%
4-Methyl-2-nitrophenyl isothiocyanate
Formula: C₈H₆N₂O₂S
MW: 194.21 g/mol
Melting Pt: 62…67 °C
Storage Temperature: Ambient
MDL Number: MFCD00041248
CAS Number: 17614-74-3
EINECS: 000-000-0
UN: 2923
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR