5-(2,6-Dichlorophenyl)-1H-tetrazole ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 5-(2-Fluorophenyl)tetrazole

L19258.MD
L19258.MDEA 34.5 EUR
L19258.MD
5-(2,6-Dichlorophenyl)-1H-tetrazole ≥97%
5-(2,6-Dichlorophenyl)-1H-tetrazole
Formula: C₇H₄Cl₂N₄
MW: 215.04 g/mol
Melting Pt: 203…209 °C
Storage Temperature: Ambient
MDL Number: MFCD00798373
CAS Number: 50907-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR