3-Bromo-2-thenoic acid

Supplier: Thermo Scientific

Synonyms: 3-Bromo-2-thiophenecarboxylic acid,3-Bromothiophene-2-carboxylic acid

SEW01838DA
MAYBSEW01838DAEA 0 EUR
MAYBSEW01838DA
3-Bromo-2-thenoic acid
3-Bromo-2-thenoic acid
Formula: C₅H₃BrO₂S
MW: 207.05 g/mol
MDL Number: MFCD00052291
CAS Number: 7311-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR