Trimethylsulphonium iodide

Supplier: EHRENSTORFER GMBH
Warning

C17885000
EHERC17885000EA 74.4 EUR
EHERC17885000
Trimethylsulphonium iodide
Trimethylsulphonium iodide
Formula: C₃H₉IS
MW: 204.07 g/mol
Melting Pt: 215…220 °C
Density: 1,922 g/cm³ (20 °C)
MDL Number: MFCD00011632
CAS Number: 2181-42-2
EINECS: 218-555-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR