(4-Chlorophenoxy)acetic acid, Supelco®

Supplier: Merck
Warning

Synonyms: 4-CPA

M-Clarity™ quality level = MQ200

45391-250MG
45391-250MGEA 19 EUR
45391-250MG
(4-Chlorophenoxy)acetic acid, Supelco®
(4-Chlorophenoxy)acetic acid
Formula: (C₆H₄Cl)OCH₂COOH
MW: 186.59 g/mol
Boiling Pt: 308 °C (1013 hPa)
Melting Pt: 160 °C
Density: 1.53 g/cm³ (20 °C)
MDL Number: MFCD00004305
CAS Number: 122-88-3
EINECS: 204-581-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR