Sodium acetate 0.1 mol/l (0.1 N) in denatured ethanol 96%
Supplier: Bernd Kraft
Synonyms: Acetic acid sodium salt
16590.3700 16590.2700 16590.4700
KRAF16590.3700EA 95 EUR
KRAF16590.3700 KRAF16590.2700 KRAF16590.4700
Sodium acetate 0.1 mol/l (0.1 N) in denatured ethanol 96%
Sodium acetate
Formula: H₃CCOONa
MW: 82.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012459
CAS Number: 127-09-3
EINECS: 204-823-8