1-Butyl-3-methylimidazoliumhexafluorophosphate, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 1-Butyl-3-methyl-1H-imidazol-3-ium hexafluorophosphate(V),1-(But-1-yl)-3-methyl-1H-imidazol-3-ium hexafluorophosphate
M-Clarity™ quality level = MQ200
4.90050.0500 4.90050.1000
4.90050.0500EA 2790 EUR
4.90050.0500 4.90050.1000
1-Butyl-3-methylimidazoliumhexafluorophosphate, Sigma-Aldrich®
1-Butyl-3-methylimidazolium hexafluorophosphate
Formula: C₈H₁₅F₆N₂P
MW: 284.18 g/mol
Boiling Pt: ∼200 °C (1013 hPa)
Melting Pt: ∼6 °C
Density: 1.38 g/cm³ (20 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD03093295
CAS Number: 174501-64-5
Order Now
Specification Test Results
Melting Point 12 °C
Molar Mass 284.18 g/mol
Density 1.38 g/cm3 (20 °C)
pH 5 (H2O, 20 °C)